1-(Dimethylethylsilyl)imidazole ≥98.0% (by titrimetric analysis) dimethylethylsilylating agent
Proveedor: TCI
|
Atención
|
D1516-1G
D1516-5G
TCIAD1516-1GEA
64
EUR
TCIAD1516-1G
TCIAD1516-5G
1-(Dimethylethylsilyl)imidazole ≥98.0% (by titrimetric analysis) dimethylethylsilylating agent
1-(Dimethylethylsilyl)imidazole
Formula:
C₇H₁₄N₂Si Pto. Ebullición: 80°C |
Número MDL:
MFCD00059723 Núm. CAS: 62365-34-8 EINECS: 263-525-6 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (Nonaqueous Titration) | min. 98.0 % |
Effect test of GC derivalization | Effective as derivatization agent Estradiol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...