4-(3,5-Dibromo-2-pyridylazo)-1,3-phenylenediamine ≥98.0% (by titrimetric analysis) for colorimetric analysis of Co, Cd
Proveedor: TCI
|
Atención
|
D1552-1G
D1552-100MG
TCIAD1552-1GEA
206
EUR
TCIAD1552-1G
TCIAD1552-100MG
4-(3,5-Dibromo-2-pyridylazo)-1,3-phenylenediamine ≥98.0% (by titrimetric analysis) for colorimetric analysis of Co, Cd
4-(3,5-Dibromo-2-pyridylazo)-1,3-phenylenediamine
Formula:
C₁₁H₉Br₂N₅ |
Número MDL:
MFCD00059780 Núm. CAS: 50768-79-1 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Suitability for cobalt-analysis | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...