Dibutyl-N,N-diethylcarbamoylmethylphosphonate for extraction of lanthanides and actinides

Proveedor: TCI

Sinónimos: N,N-Diethylcarbamoylmethylphosphonic Acid Dibutyl Ester

D1585-25ML D1585-5ML
TCIAD1585-25MLEA 162 EUR
TCIAD1585-25ML TCIAD1585-5ML
Dibutyl-N,N-diethylcarbamoylmethylphosphonate for extraction of lanthanides and actinides
Dibutyl-N,N-diethylcarbamoylmethylphosphonate
Formula: C₁₄H₃₀NO₄P
MW: 307,37 g/mol
Pto. Ebullición: 172°C
Número MDL: MFCD00015240
Núm. CAS: 7439-68-1

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR