N,N-Dimethylmethyleneammonium iodide ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: N,N-Dimethylmethyleneiminium Iodide, Eschenmoser's Salt

D1646-25G D1646-5G
TCIAD1646-25GEA 158 EUR
TCIAD1646-25G TCIAD1646-5G
N,N-Dimethylmethyleneammonium iodide ≥97.0% (by titrimetric analysis)
N,N-Dimethylmethyleneammonium iodide
Formula: C₃H₈IN
MW: 185,01 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00011810
Núm. CAS: 33797-51-2
EINECS: 251-680-2
Índice Merck: 14,03251

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (Potassium iodate Method) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR