4-Dimethylamino-1-naphthyl isothiocyanate ≥98.0% (by HPLC, titration analysis) for HPLC labeling
Proveedor: TCI
|
Atención
|
Sinónimos:
DANITO
D1669-1G
D1669-5G
TCIAD1669-1GEA
88
EUR
TCIAD1669-1G
TCIAD1669-5G
4-Dimethylamino-1-naphthyl isothiocyanate ≥98.0% (by HPLC, titration analysis) for HPLC labeling
4-Dimethylamino-1-naphthyl isothiocyanate
Formula:
C₁₃H₁₂N₂S MW: 228,31 g/mol |
Número MDL:
MFCD00003883 Núm. CAS: 29711-79-3 EINECS: 249-800-3 |
Resultados test especificaciones
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 67.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...