2,3-Diphenylpropionic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

D1790-25G D1790-5G
TCIAD1790-25GEA 366 EUR
TCIAD1790-25G TCIAD1790-5G
2,3-Diphenylpropionic acid ≥98.0% (by GC, titration analysis)
2,3-Diphenylpropionic acid
Formula: C₁₅H₁₄O₂
MW: 226,28 g/mol
Pto. Ebullición: 340°C
Número MDL: MFCD00044405
Núm. CAS: 3333-15-1
EINECS: 222-065-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR