3,6-Dihydroxybenzonorbornane ≥98.0% (by HPLC, titration analysis) antioxidant
Proveedor: TCI
Sinónimos:
Benzonorbornene-3,6-diol, 1,2,3,4-Tetrahydro-1,4-methanonaphthalene-5,8-diol
D1806-25G
D1806-5G
TCIAD1806-25GEA
326
EUR
TCIAD1806-25G
TCIAD1806-5G
3,6-Dihydroxybenzonorbornane ≥98.0% (by HPLC, titration analysis) antioxidant
3,6-Dihydroxybenzonorbornane
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol |
Número MDL:
MFCD00077441 Núm. CAS: 16144-91-5 EINECS: 240-300-0 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Cerium redox Method) | min. 98.0 % |
Melting point | 177.0 to 181.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...