3,5-Di-tert-butylsalicylic acid ≥95.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
D1947-25G
D1947-500G
TCIAD1947-25GEA
27
EUR
TCIAD1947-25G
TCIAD1947-500G
3,5-Di-tert-butylsalicylic acid ≥95.0% (by titrimetric analysis)
3,5-Di-tert-butylsalicylic acid
Formula:
C₁₅H₂₂O₃ MW: 250,34 g/mol |
Número MDL:
MFCD00059058 Núm. CAS: 19715-19-6 EINECS: 243-246-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystaline |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 159.0 to 166.0 deg-C |
Solubility in Methanol | very faint turbidity |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...