S,S-Diphenylsulphilimine monohydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
S,S-Diphenylsulphilimine monohydrate, Sulphilimine, S,S-diphenyl-, monohydrate, S,S-Diphenylsulphimine monohydrate
D2002-1G
D2002-5G
TCIAD2002-1GEA
98
EUR
TCIAD2002-1G
TCIAD2002-5G
S,S-Diphenylsulphilimine monohydrate ≥98.0% (by titrimetric analysis)
S,S-Diphenylsulphilimine monohydrate
Formula:
C₁₂H₁₁NS MW: 201,29 g/mol |
Número MDL:
MFCD00001757 Núm. CAS: 68837-61-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0% (calcd.on anh.substance) |
Melting point | 71.0 to 75.0 deg-C |
Solubility in Methanol | within almost transparency |
Water | 7.5 to 9.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...