Sodium diphenylphosphinobenzene-3-sulphonate ≥90.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Sodium 3-(diphenylphosphino)benzenesulphonate, Diphenylphosphinobenzene-3-sulphonic acid sodium salt, Triphenylphosphine-3-sulphonic acid sodium salt
D2043-1G
D2043-5G
TCIAD2043-1GEA
67
EUR
TCIAD2043-1G
TCIAD2043-5G
Sodium diphenylphosphinobenzene-3-sulphonate ≥90.0% (by titrimetric analysis)
Sodium diphenylphosphinobenzene-3-sulphonate
Formula:
C₁₈H₁₅O₃PS MW: 364,33 g/mol |
Número MDL:
MFCD00144157 Núm. CAS: 63995-75-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...