1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers) ≥97.0% (por GC)

Proveedor: TCI

D2077-1G D2077-5G
TCIAD2077-1GEA 106 EUR
TCIAD2077-1G TCIAD2077-5G
1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers) ≥97.0% (por GC)
1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers)
Formula: C₁₂H₁₀S₂
MW: 218.33 g/mol
Número MDL: MFCD00142549
Núm. CAS: 23354-93-0

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR