4,4'-Oxybis(benzoic acid) ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4′-Oxybis(benzoic acid)
D2115-25G
D2115-100G
TCIAD2115-25GEA
26
EUR
TCIAD2115-25G
TCIAD2115-100G
4,4'-Oxybis(benzoic acid) ≥98.0% (by GC, titration analysis)
4,4'-Oxybis(benzoic acid)
Formula:
C₁₄H₁₀O₅ MW: 258,23 g/mol |
Número MDL:
MFCD00013988 Núm. CAS: 2215-89-6 EINECS: 218-683-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in 1mol/L NaOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...