3,4-Diisopropoxy-3-cyclobutene-1,2-dione ≥98.0% (por GC)

Proveedor: TCI
Peligro

Sinónimos: Diisopropyl Squarate, 3,4-Diisopropoxycyclobut-3-ene-1,2-dione

D2207-25G D2207-5G
TCIAD2207-25GEA 232 EUR
TCIAD2207-25G TCIAD2207-5G
3,4-Diisopropoxy-3-cyclobutene-1,2-dione ≥98.0% (por GC)
3,4-Diisopropoxy-3-cyclobutene-1,2-dione
Formula: C₁₀H₁₄O₄
MW: 198,22 g/mol
Número MDL: MFCD00075125
Núm. CAS: 61699-62-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR