Dipyridamole ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 2,6-Bis(diethanolamine)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, 2,2',2'',2'''-((4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl)bis(azanetriyl))tetrakis(ethan-1-ol), 2,2',2'',2'''-[(4,8-Dipiperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakisethanol

D2274-25G D2274-5G
TCIAD2274-25GEA 95 EUR
TCIAD2274-25G TCIAD2274-5G
Dipyridamole ≥98.0% (by HPLC, titration analysis)
Dipyridamole
Formula: C₂₄H₄₀N₈O₄
MW: 504.64 g/mol
Número MDL: MFCD00010555
Núm. CAS: 58-32-2
EINECS: 200-374-7
Índice Merck: 14,03346

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in THF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR