Diphenyl-2-pyridylphosphine ≥97.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(Diphenylphosphino)pyridine
D2471-1G
D2471-5G
TCIAD2471-1GEA
18
EUR
TCIAD2471-1G
TCIAD2471-5G
Diphenyl-2-pyridylphosphine ≥97.0% (por GC)
Diphenyl-2-pyridylphosphine
Formula:
C₁₇H₁₄NP MW: 263,28 g/mol |
Número MDL:
MFCD00192108 Núm. CAS: 37943-90-1 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 84.0 to 87.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...