Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Di-μ-hydroxo-bis[(N,N,N'.N'-tetramethylethylenediamine)copper(II)] chloride
D2542-25G
D2542-5G
TCIAD2542-25GEA
171
EUR
TCIAD2542-25G
TCIAD2542-5G
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride ≥97.0% (by titrimetric analysis)
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride
Formula:
C₁₂H₃₄Cl₂Cu₂N₄O₂ MW: 464,42 g/mol |
Número MDL:
MFCD01321163 Núm. CAS: 30698-64-7 |
Resultados test especificaciones
Appearance | Light gray to Brown to Black powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...