Dodecanedioyl dichloride ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
D2544-25G
D2544-5G
TCIAD2544-25GEA
133
EUR
TCIAD2544-25G
TCIAD2544-5G
Dodecanedioyl dichloride ≥98.0% (by GC, titration analysis)
Dodecanedioyl dichloride
Formula:
C₁₂H₂₀Cl₂O₂ MW: 267.19 g/mol |
Número MDL:
MFCD00012243 Núm. CAS: 4834-98-4 UN: 3265 ADR: 8 III |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...