2,9-Dibutyl-1,10-phenanthroline ≥98.0% (by total nitrogen basis)

Proveedor: TCI

D2565-1G D2565-100MG
TCIAD2565-1GEA 331 EUR
TCIAD2565-1G TCIAD2565-100MG
2,9-Dibutyl-1,10-phenanthroline ≥98.0% (by total nitrogen basis)
2,9-Dibutyl-1,10-phenanthroline
Formula: C₂₀H₂₄N₂
MW: 292,43 g/mol
Número MDL: MFCD02093459
Núm. CAS: 85575-93-5

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 57.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR