N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥96.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: N,N'-Bis(2-hydroxybenzylidene)-1,2-phenylenediamine, N,N'-Disalicylal-1,2-phenylenediamine, 2,2'-((1,2-Phenylenebis(azanylylidene))bis(methanylylidene))diphenol

D2572-25G D2572-5G
TCIAD2572-25GEA 218 EUR
TCIAD2572-25G TCIAD2572-5G
N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥96.0% (by HPLC, total nitrogen)
N,N'-Bis(salicylidene)-1,2-phenylenediamine
Formula: C₂₀H₁₆N₂O₂
MW: 316,36 g/mol
Número MDL: MFCD00009995
Núm. CAS: 3946-91-6

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Melting point 163.0 to 166.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR