N,N-Diethylaniline borane ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Borane - N,N-Diethylaniline Complex

D2581-100G D2581-25G
TCIAD2581-100GEA 527 EUR
TCIAD2581-100G TCIAD2581-25G
N,N-Diethylaniline borane ≥98.0% (by titrimetric analysis)
N,N-Diethylaniline borane
Formula: C₁₀H₁₈BN
MW: 163.07 g/mol
Número MDL: MFCD00013187
Núm. CAS: 13289-97-9
EINECS: 236-305-2
UN: 3148
ADR: 4.3 III

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR