Methyl-3,4-dihydroxyphenylacetate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: 3,4-Dihydroxyphenylacetic Acid Methyl Ester, Methyl 2-(3,4-dihydroxyphenyl)acetate, Methyl 3,4-dihydroxyphenylacetate, Catechol-4-acetic Acid Methyl Ester, Homoprotocatechuic Acid Methyl Ester, Pyrocatechol-4-acetic Acid Methyl Ester

D2734-1G D2734-5G
TCIAD2734-1GEA 69 EUR
TCIAD2734-1G TCIAD2734-5G
Methyl-3,4-dihydroxyphenylacetate ≥98.0% (por GC)
Methyl-3,4-dihydroxyphenylacetate
Formula: C₉H₁₀O₄
MW: 182,18 g/mol
Número MDL: MFCD00583539
Núm. CAS: 25379-88-8

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR