4,4'-Diiodo-3,3'-dimethylbiphenyl ≥98.0% (por GC)
Proveedor: TCI
D2759-25G
D2759-5G
TCIAD2759-25GEA
650
EUR
TCIAD2759-25G
TCIAD2759-5G
4,4'-Diiodo-3,3'-dimethylbiphenyl ≥98.0% (por GC)
4,4'-Diiodo-3,3'-dimethylbiphenyl
Formula:
C₁₄H₁₂I₂ |
Número MDL:
MFCD04038414 Núm. CAS: 7583-27-9 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...