Ethyl-5,6-dichloronicotinate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: 5,6-Dichloronicotinic Acid Ethyl Ester, 5,6-Dichloropyridine-3-carboxylic Acid Ethyl Ester, Ethyl 5,6-dichloronicotinate, Ethyl 5,6-Dichloropyridine-3-carboxylate

D2772-25G D2772-5G
TCIAD2772-25GEA 425 EUR
TCIAD2772-25G TCIAD2772-5G
Ethyl-5,6-dichloronicotinate ≥98.0% (por GC)
Ethyl-5,6-dichloronicotinate
Formula: C₈H₇Cl₂NO₂
MW: 220,05 g/mol
Número MDL: MFCD03093627
Núm. CAS: 401566-69-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR