4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

Sinónimos: 2-(2,6-Dimethyl-4H-pyran-4-ylidene)propanedinitrile, 2-(2,6-Dimethyl-4H-pyran-4-ylidene)malononitrile

D2791-1G D2791-5G
TCIAD2791-1GEA 70 EUR
TCIAD2791-1G TCIAD2791-5G
4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran ≥98.0% (by HPLC, total nitrogen)
4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran
Formula: C₁₀H₈N₂O
MW: 172,19 g/mol
Número MDL: MFCD00181519
Núm. CAS: 28286-88-6
UN: 3439
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 192.0 to 196.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR