Dimethyl dodecanedioate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Dodecanedioic Acid Dimethyl Ester
D2835-25G
D2835-500G
TCIAD2835-25GEA
17
EUR
TCIAD2835-25G
TCIAD2835-500G
Dimethyl dodecanedioate ≥98.0% (por GC)
Dimethyl dodecanedioate
Formula:
C₁₄H₂₆O₄ MW: 258,36 g/mol |
Número MDL:
MFCD00043632 Núm. CAS: 1731-79-9 EINECS: 217-050-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...