2,2-Dibromo-2-cyanoacetamide ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
2-Cyano-2,2-dibromoacetamide
D2902-25G
D2902-500G
TCIAD2902-25GEA
23
EUR
TCIAD2902-25G
TCIAD2902-500G
2,2-Dibromo-2-cyanoacetamide ≥98.0% (by total nitrogen basis)
2,2-Dibromo-2-cyanoacetamide
Formula:
C₃H₂Br₂N₂O MW: 241,87 g/mol |
Número MDL:
MFCD00129791 Núm. CAS: 10222-01-2 EINECS: 233-539-7 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...