Dicyclohexyl(trifluoromethanesulphonyloxy)borane ≥98.0% (by NMR)

Proveedor: TCI
Peligro

Sinónimos: Trifluoromethanesulphonic acid Dicyclohexylboryl Ester, Dicyclohexylboron Triflate

D2943-1G D2943-5G
TCIAD2943-1GEA 52 EUR
TCIAD2943-1G TCIAD2943-5G
Dicyclohexyl(trifluoromethanesulphonyloxy)borane ≥98.0% (by NMR)
Dicyclohexyl(trifluoromethanesulphonyloxy)borane
Formula: C₁₃H₂₂BF₃O₃S
MW: 326,18 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD06797094
Núm. CAS: 145412-54-0
UN: 1759
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (NMR) min. 98.0 atom%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR