N,N'-Dimethoxy-N,N'-dimethyloxamide ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
N1,N2-Dimethoxy-N1,N2-dimethyloxalamide, N,N'-Oxalylbis[methoxy(methyl)amine]
D2961-25G
D2961-5G
TCIAD2961-25GEA
387
EUR
TCIAD2961-25G
TCIAD2961-5G
N,N'-Dimethoxy-N,N'-dimethyloxamide ≥98.0% (por GC)
N,N'-Dimethoxy-N,N'-dimethyloxamide
Formula:
C₆H₁₂N₂O₄ MW: 176,17 g/mol |
Número MDL:
MFCD02326951 Núm. CAS: 106675-70-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...