N,N-Dibenzylglycine ethyl ester ≥98.0% (by GC, total nitrogen)
Proveedor: TCI
Sinónimos:
N,N-Dibenzylglycine Ethyl Ester, Di-Bzl-Gly-OEt, Ethyl 2-(dibenzylamino)acetate
D3165-25G
D3165-5G
TCIAD3165-25GEA
270
EUR
TCIAD3165-25G
TCIAD3165-5G
N,N-Dibenzylglycine ethyl ester ≥98.0% (by GC, total nitrogen)
N,N-Dibenzylglycine ethyl ester
Formula:
C₁₈H₂₁NO₂ MW: 283,37 g/mol |
Número MDL:
MFCD00796343 Núm. CAS: 77385-90-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...