1,8-Dibromopyrene ≥97.0% (por GC)
Proveedor: TCI
D3168-1G
D3168-200MG
TCIAD3168-1GEA
338
EUR
TCIAD3168-1G
TCIAD3168-200MG
1,8-Dibromopyrene ≥97.0% (por GC)
1,8-Dibromopyrene
Formula:
C₁₆H₈Br₂ MW: 360,05 g/mol |
Número MDL:
MFCD00230868 Núm. CAS: 38303-35-4 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 209.0 to 213.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...