7-(Diethylamino)coumarin ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
7-(Diethylamino)-2H-chromen-2-one
D3354-200MG
D3354-1G
TCIAD3354-200MGEA
35
EUR
TCIAD3354-200MG
TCIAD3354-1G
7-(Diethylamino)coumarin ≥98.0% (by GC, titration analysis)
7-(Diethylamino)coumarin
Formula:
C₁₃H₁₅NO₂ MW: 217,27 g/mol |
Número MDL:
MFCD00232946 Núm. CAS: 20571-42-0 EINECS: 243-888-7 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...