3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
D3452-5G
D3452-25G
TCIAD3452-5GEA
17
EUR
TCIAD3452-5G
TCIAD3452-25G
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid
Formula:
C₁₇H₂₆O₃ MW: 278,40 g/mol |
Número MDL:
MFCD00017519 Núm. CAS: 20170-32-5 EINECS: 243-556-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...