N,N'-Di-p-tolylbenzidine ≥98.0% (por GC)

Proveedor: TCI

D3567-1G D3567-5G
TCIAD3567-1GEA 45 EUR
TCIAD3567-1G TCIAD3567-5G
N,N'-Di-p-tolylbenzidine ≥98.0% (por GC)
N,N'-Di-p-tolylbenzidine
Formula: C₂₆H₂₄N₂
Número MDL: MFCD09833311
Núm. CAS: 10311-61-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 237.0 to 243.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR