9,9-Diphenylfluorene ≥98.0% (por GC)
Proveedor: TCI
D3586-5G
D3586-25G
TCIAD3586-5GEA
60
EUR
TCIAD3586-5G
TCIAD3586-25G
9,9-Diphenylfluorene ≥98.0% (por GC)
9,9-Diphenylfluorene
Formula:
C₂₅H₁₈ |
Número MDL:
MFCD00093944 Núm. CAS: 20302-14-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 224.0 to 228.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...