3,3'-Dibromo-4,4'-biphenol ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 3,3'-Dibromo-4,4'-dihydroxybiphenyl, 3,3'-Dibromobiphenyl-4,4'-diol, 3,3'-Dibromo-[1,1'-biphenyl]-4,4'-diol

D3602-1G D3602-5G
TCIAD3602-1GEA 172 EUR
TCIAD3602-1G TCIAD3602-5G
3,3'-Dibromo-4,4'-biphenol ≥98.0% (by titrimetric analysis)
3,3'-Dibromo-4,4'-biphenol
Formula: C₁₂H₈Br₂O₂
MW: 344,00 g/mol
Número MDL: MFCD10566892
Núm. CAS: 189039-64-3

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR