2'-Deoxy-2'-fluorouridine ≥97.0% (by HPLC)
Proveedor: TCI
Sinónimos:
2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
D3615-1G
D3615-5G
TCIAD3615-1GEA
36
EUR
TCIAD3615-1G
TCIAD3615-5G
2'-Deoxy-2'-fluorouridine ≥97.0% (by HPLC)
2'-Deoxy-2'-fluorouridine
Formula:
C₉H₁₁FN₂O₅ MW: 246,19 g/mol |
Número MDL:
MFCD01317293 Núm. CAS: 784-71-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 147.0 to 153.0 deg-C |
Specific rotation [a]20/D | +50.0 to +56.0 deg (C=0.3, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...