1,3-Diisopropylimidazolinium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

Sinónimos: IiPrim.HBF4

D3622-1G D3622-5G
TCIAD3622-1GEA 164 EUR
TCIAD3622-1G TCIAD3622-5G
1,3-Diisopropylimidazolinium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1,3-Diisopropylimidazolinium tetrafluoroborate
Formula: C₉H₁₉BF₄N₂
MW: 242,07 g/mol
Número MDL: MFCD08276803
Núm. CAS: 137581-18-1
UN: 1759
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR