1,2-Dimethylbenzimidazole ≥98.0% (by GC, titration analysis)

Proveedor: TCI

D3665-1G D3665-5G
TCIAD3665-1GEA 109 EUR
TCIAD3665-1G TCIAD3665-5G
1,2-Dimethylbenzimidazole ≥98.0% (by GC, titration analysis)
1,2-Dimethylbenzimidazole
Formula: C₉H₁₀N₂
Pto. Ebullición: 164°C
Número MDL: MFCD00022840
Núm. CAS: 2876-08-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR