4,6-Dimethyl-2-methylsulphonylpyrimidine ≥98.0% (por GC)
Proveedor: TCI
D3699-25G
D3699-5G
TCIAD3699-25GEA
211
EUR
TCIAD3699-25G
TCIAD3699-5G
4,6-Dimethyl-2-methylsulphonylpyrimidine ≥98.0% (por GC)
4,6-Dimethyl-2-methylsulphonylpyrimidine
Formula:
C₇H₁₀N₂O₂S MW: 186,23 g/mol |
Número MDL:
MFCD03788286 Núm. CAS: 35144-22-0 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 85.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...