Diphenyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate ≥95.0% (por GC)
Proveedor: TCI
Sinónimos:
(N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diphenyl Ester
D3709-1G
D3709-5G
TCIAD3709-1GEA
110
EUR
TCIAD3709-1G
TCIAD3709-5G
Diphenyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate ≥95.0% (por GC)
Diphenyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate
Formula:
C₁₆H₁₈NO₅P MW: 335,30 g/mol |
Núm. CAS:
367508-01-8 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...