Dibenzo[g,p]chrysene ≥98.0% (by HPLC)

Proveedor: TCI

D3736-1G D3736-200MG
TCIAD3736-1GEA 314 EUR
TCIAD3736-1G TCIAD3736-200MG
Dibenzo[g,p]chrysene ≥98.0% (by HPLC)
Dibenzo[g,p]chrysene
Formula: C₂₆H₁₆
MW: 328,41 g/mol
Núm. CAS: 191-68-4
EINECS: 205-890-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 215.0 to 221.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR