Diazolidinyl urea ≥97.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea
D3769-25G
D3769-250G
TCIAD3769-25GEA
24
EUR
TCIAD3769-25G
TCIAD3769-250G
Diazolidinyl urea ≥97.0% (by total nitrogen basis)
Diazolidinyl urea
Formula:
C₈H₁₄N₄O₇ MW: 278,22 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD03547942 Núm. CAS: 78491-02-8 EINECS: 278-928-2 Índice Merck: 14,03000 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...