N,N'-Dicyclohexylmorpholine-4-carboximidamide ≥98.0% (by GC, titration analysis)

Proveedor: TCI

D3792-25G D3792-5G
TCIAD3792-25GEA 51 EUR
TCIAD3792-25G TCIAD3792-5G
N,N'-Dicyclohexylmorpholine-4-carboximidamide ≥98.0% (by GC, titration analysis)
N,N'-Dicyclohexylmorpholine-4-carboximidamide
Formula: C₁₇H₃₁N₃O
MW: 293,45 g/mol
Número MDL: MFCD00063252
Núm. CAS: 4975-73-9
EINECS: 225-622-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4054fd8aa2168862b00a3d91e3e8d4a4f0eda2c8&vid=4054fd8aa2168862b00a3d91e3e8d4a4f0eda2c8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR