Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) dimer ≥95.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Najera Catalyst I
D3806-1G
D3806-250MG
TCIAD3806-1GEA
260
EUR
TCIAD3806-1G
TCIAD3806-250MG
Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) dimer ≥95.0% (by titrimetric analysis)
Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) dimer
Formula:
C₂₆H₁₆Cl₆N₂O₂Pd₂ MW: 813,97 g/mol |
Número MDL:
MFCD09038424 Núm. CAS: 287410-78-0 |
Resultados test especificaciones
Appearance | White to Amber powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...