3-(Dodecylthio)propionic acid ≥90.0% (por GC)

Proveedor: TCI
Atención

D3814-25G D3814-500G
TCIAD3814-25GEA 32 EUR
TCIAD3814-25G TCIAD3814-500G
3-(Dodecylthio)propionic acid ≥90.0% (por GC)
3-(Dodecylthio)propionic acid
Formula: C₁₅H₃₀O₂S
Núm. CAS: 1462-52-8
EINECS: 215-967-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR