1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(2-(Di-tert-butylphosphino)phenyl)-3,5-diphenyl-1H-pyrazole
D3834-1G
D3834-5G
TCIAD3834-1GEA
53
EUR
TCIAD3834-1G
TCIAD3834-5G
1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole
Formula:
C₂₉H₃₃N₂P MW: 440,57 g/mol |
Número MDL:
MFCD09038823 Núm. CAS: 628333-86-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...