(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(R)-(+)-2-[Diphenyl(trimethylsilyloxy)methyl]pyrrolidine, (R)-Hayashi-Jorgensen Catalyst, alpha,alpha-Diphenyl-D-prolinol trimethylsilyl ether
D3867-1G
D3867-5G
TCIAD3867-1GEA
195
EUR
TCIAD3867-1G
TCIAD3867-5G
(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)
(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether
Formula:
C₂₀H₂₇NOSi MW: 325,53 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD09265100 Núm. CAS: 943757-71-9 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +53.0 to +57.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...