Dichloro(1,10-phenanthroline)copper(II) ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: (1,10-Phenanthroline)copper(II) Dichloride, Dichloro(1,10-phenanthroline)copper

D3891-1G D3891-5G
TCIAD3891-1GEA 31 EUR
TCIAD3891-1G TCIAD3891-5G
Dichloro(1,10-phenanthroline)copper(II) ≥98.0% (by total nitrogen and titration analysis)
Dichloro(1,10-phenanthroline)copper(II)
Formula: C₁₂H₈Cl₂CuN₂
MW: 314,66 g/mol
Número MDL: MFCD00134224
Núm. CAS: 14783-09-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR