1-Benzhydrylazetidin-3-ol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-(Diphenylmethyl)azetidin-3-ol

D3907-25G D3907-5G
TCIAD3907-25GEA 151 EUR
TCIAD3907-25G TCIAD3907-5G
1-Benzhydrylazetidin-3-ol ≥98.0% (by GC, titration analysis)
1-Benzhydrylazetidin-3-ol
Formula: C₁₆H₁₇NO
MW: 239.32 g/mol
Número MDL: MFCD00205109
Núm. CAS: 18621-17-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR