9,9-Dimethylfluoren-2-boronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (9,9-Dimethyl-9H-fluoren-2-yl)boronic acid

D3974-1G D3974-5G
TCIAD3974-1GEA 27 EUR
TCIAD3974-1G TCIAD3974-5G
9,9-Dimethylfluoren-2-boronic acid (contains varying amounts of Anhydride)
9,9-Dimethylfluoren-2-boronic acid
Formula: C₁₅H₁₅BO₂
MW: 238,09 g/mol
Número MDL: MFCD08704227
Núm. CAS: 333432-28-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR